Would you like to log in again?
CPEP Expanding Universe Activity
Supplier: Avantor
WARD470199-820
470199-820EA 99.99 USD InStock
470199-820
CPEP Expanding Universe Activity
Educational Materials Physics Educational Materials Modern Physics Learning Activities
Explain the complex concepts of the expanding universe using the simple models of this two-part teacher demonstration and student lab. The demonstration models how light waves are stretched to greater wavelengths as space expands, while the lab activity allows students to visualize the distance of galaxies, as seen by an observer, by simulating the direct relationship between a galaxy’s distance from the observer and its expansion velocity.
Developed by the Contemporary Physics Education Project (CPEP) and aligned to national standards, the set up and procedure for each activity is displayed on the included instructional DVD. Includes materials for 10 lab groups. Grades 9 - 12.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty