Would you like to log in again?
Crossing Over in Sordaria Slide
Supplier: Avantor
Avg. Ratings: 4.5 (2 Reviews)
REVIEWS (2)
Total Ratings: 2
Avg. Ratings: 4.5 out of 5
Sort all by:
Was this review helpful? Yes No
Yes: 0 No: 0 
Education / AP Biology Teacher / Lexington, KY
Yes, I recommend
I love having these slides as back up if my students have difficulty using the live Sordaria Plate or I don't order it in time for the lab. Every slide has evidence of crossing over and students can easy measure the cross over frequency.
Reply
vwr.com associate Published 11/14/2017
We're thrilled to hear you are enjoying our Crossing Over in Sordaria. Thanks for sharing!
Was this review helpful? Yes No
Yes: 3 No: 0 
Rochester, NY
Yes, I recommend
Great slide! - When we tried to make our own slides with Sordaria, we were unsuccessful, but these slides allow us to show real results clearly and effectively!
WARD470177-436
470177-436EA 26.25 USD InStock
470177-436
Crossing Over in Sordaria Slide
Slides Prepared Slides
Whole mount
  • All identifying features are clearly distinguishable
  • Rigorous quality control standards
Shows a wild-type and a mutant cross. The arrangement of ascospores in the asci are clearly displayed and are used to estimate the percentage of crossing over between the centromere and the gene that controls the tan spore color.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (2)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty