Would you like to log in again?
Cyanobacteria Morphology Set
Supplier: VWR
WARD470179-928
470179-928KT 36 USD InStock
470179-928
Cyanobacteria Morphology Set
Specimens Live Specimens Bacteria - Living Material
Study Structural Variety in Blue-Green Algae
This collection of different types of cyanobacteria displays their structural diversity. Cultures: Oscillatoria, Nostoc (filamentous); Gloeocapsa (colonial globular); and Gloeotrichia (colonial filamentous). The set of four includes the Working with Algae manual and four pipets.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty