Would you like to log in again?
Cylindrical Metal Mirror
Supplier: STEINER ENTERPRISES SE
3524
470111-312EA 18.19 USD InStock
470111-312
Cylindrical Metal Mirror
Mirrors
Show Reflection and Refraction
  • Heavily plated
A concave/convex cylindrical metal mirror, heavily plated, is polished on both sides. A 7.5 x 5cm plate has a 5cm radius of curvature. Comes with protective plastic sheets on both surfaces, to be removed before use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty