Would you like to log in again?
Ward's® Live Cylindrospermum Culture
Supplier: VWR
WARD470179-866 WARD470179-868
470179-868EA 29.95 USD InStock
470179-868 470179-866
Ward's® Live Cylindrospermum Culture
Specimens Live Specimens Bacteria - Living Material
This filamentous cyanobacteria is comprised of heterocysts and akinetes that are terminally located on the filament.
Commonly found in both terrestrial and aquatic ecosystems,
Genus: Cylindrospermum Species: Not specified
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty