Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DC Motor

Supplier: VWR International
PH1242
470111-202EA 29.85 USD InStock
470111-202
DC Motor
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This DC Motor arrives completely assembled.

  • Storage Case Included
  • Open Design
  • Reversing Magnets Changes the Direction of Motor's Rotation
  • Motor Can Be Used Vertically or Horizontally
  • For Experiments and Demonstrations

This open design is similar to diagrams in most textbooks and makes it easy to study motor-related principles. Users can reverse magnets to change direction of the motor’s rotation. Users can also observe change of speed with current with an external rheostat (not included).

The lightweight motor can be used vertically or horizontally for experiments and demonstrations. It can be used with a 1.5V battery or external, user-provided power supply.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty