Would you like to log in again?
Ward's® Chemistry Dehydration of Sucrose Demonstration
Supplier: Avantor
WARD470010-820
470010-820EA 58.99 USD InStock
470010-820
Ward's® Chemistry Dehydration of Sucrose Demonstration
Educational Classroom Kits and Activities
Amaze Students with Chemical Phenomena
  • Perfect supplement to your chemistry lectures
  • Can be performed in just 10–30 minutes
  • Requires only basic labware that you provide
In this demonstration you will pour sulfuric acid on a mixture of sucrose and sodium carbonate to produce a large amount of black foam, steam and smoke.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty