Would you like to log in again?
Compact Electrophoresis Power Supply
Supplier: Avantor
105ECA-115
470204-250EA 1229.8 USD InStock
470204-250
Compact Electrophoresis Power Supply
Power Supplies
This compact power supply is ideal for DNA or RNA electrophoresis.
  • Easy-to-use power supply operates in constant voltage
  • Adjustment knob and hi-lo range switch control the output, which is variable in 10 V increments from 20 V to 250 V
  • Two-position switch toggles the bright LED display from indicating output voltage to output current
  • Equipped with an overcurrent detector for maximum safety
Carry out DNA and RNA electrophoresis with the help of the Owl EC-105 Compact Power Supply, equipped with an overcurrent detector for maximum safety. Unit has two jacks to run two gel boxes simultaneously.
Delivery information: This item ships directly from our supplier.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty