Would you like to log in again?
Demo Electric Motor
Supplier: VWR International
PH1237
470014-464EA 79.45 USD InStock
470014-464
Demo Electric Motor
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The construction of the multi-functional Demo Electric Motor is completely open.
  • Three Copper Wire-Wound Armatures
  • Visible Parts
  • Removable Bar Magnet Produces a Permanent Magnetic Field
  • Phosphor-Bronze Brushes
  • Motor Operates on 4.5—9VDC
All parts of the motor are visible, so students can observe their inner workings. A model of a simple DC electric motor, this device has three armatures: two pole, three pole, and four pole, which are wound with enameled copper wire. A removable bar magnet produces a permanent magnetic field.
A disc-type commentator is incorporated, and a pair of 4 mm sockets provides an external connection to the phosphor-bronze brushes. The motor operates on 4.5—9VDC.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty