Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Denoyer-Geppert® Cardiopulmonary System
Supplier: DENOYER-GEPPERT SCI CO SE
0192-00
470004-198EA 879.8 USD InStock
470004-198
Denoyer-Geppert® Cardiopulmonary System
Models Human Biology Models
This system shows interdependence of circulatory and respiratory systems.
  • Wooden Base Mount
  • Removable Parts
  • Detailed Depictions
  • Sectioned Larynx
The life-size heart and lungs are depicted in great detail and vivid color to display all the important structures of both systems involved in pulmonary circulation.
Half of the left lung and the two-part heart are removable to show bifurcation of the trachea, bronchial tree, and pulmonary vessels. The larynx is sectioned, and the right half is removable for closer study. The esophagus and major vessels are also shown.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty