Would you like to log in again?
Denoyer-Geppert® Upper Abdominal Organs Model
Supplier: DENOYER-GEPPERT SCI CO SE
0146-00
470006-248EA 716.6 USD InStock
470006-248
Denoyer-Geppert® Upper Abdominal Organs Model
Models Human Biology Models
A comprehensive view of overlooked organs.
  • Three Times Larger than Life Size
  • Model Includes Detailed Nephrons and Glomeruli
  • Complete Depiction of the Digestive System with Seven Organs
The featured organs are the liver, kidneys and adrenal glands, gallbladder, pancreas, duodenum, and spleen. The gallbladder can be removed to reveal the cystic duct with spiral valve. The pancreas, duodenum, and gallbladder are all dissected to show the pancreatic, cystic, and bile ducts.
The spleen is detailed and pulled away from the left kidney to reveal the blood vessels that supply it. The hepatic portal vein and pancreatic arteries are also included. The two-piece model is mounted on a wooden base and can be removed for up-close study.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty