Would you like to log in again?
Denoyer-Geppert® Dicot Flower Model
Supplier: DENOYER-GEPPERT SCI CO SE
0824-50 0500-00
470029-524EA 393.6 USD InStock
470029-524 470006-246
Denoyer-Geppert® Dicot Flower Model
Models Botany Models
This dissectible flower helps students learn about angiosperm reproduction.
  • Model Enlarged 10 Times
  • Teacher's Guide Included
  • Key Describing 28 Structures Included
  • Eight Flower Parts in Model
Each of the eight parts of the flower model represents one of the unique structural components such as the sepals, petals, stamens, and pistil. The model shows fertilization with pollen grains on the stigma; one pollen tube is shown penetrating the micropyle. The anthers are sectioned, and the ovary is dissected to show the female gametophyte. The model also depicts the scenario for double fertilization.
The eight-piece model is mounted on a base and includes a teacher’s guide and a key identifying 28 structures. The student version incorporates fewer detachable pieces and a more simplified painting to result in a lower price.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty