Would you like to log in again?
Dictyostelium Culture and Study Kit
Supplier: VWR
470179-640KT 74.95 USD InStock
470179-640
Dictyostelium Culture and Study Kit
Specimens Live Specimens Protists - Living Material
Study Structure and Motility of a Variety of Common Protists
Convenient, all-inclusive kit contains everything you need to culture and examine different protists to discover their unique attributes. Comes with a specimen culture, culture materials, Form and Structure sheets, and instructions for 30 students.
These microbes are members of the cellular slime mold group, and are commonly found in certain types of soil, dung, and other high-bacteria habitats. A variety of experiments can be conducted with Dictyostelium, including observing its life history and investigating differentiation and taxis responses. It is also interesting to observe Dictyostelium feeding on bacteria, which are included in this kit. Additional suggested experiments include studies of migrating pseudoplasmodia and differentiation. Cultures: Dictyostelium discoideum, Enterobacter aerogenes, or Escherichia coli (food).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty