Would you like to log in again?
Dictyostelium discoideum, Tube Culture
Supplier: VWR
WARD470176-392
470176-392EA 12.95 USD InStock
470176-392
Dictyostelium discoideum, Tube Culture
Specimens Live Specimens Protists - Living Material
Cellular slime mold. Demonstrates amoeboid state, pseudoplasmodial state, and fruiting body, or sorocarp.
  • Found in woodland soil in decomposing vegetation where bacteria are abundant
  • Characterized by an amoeboid stage (lacking cell walls) and a fungus-like reproductive structure
  • Incubation temperature 25°C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty