Would you like to log in again?
Diffraction Grating
Supplier: IMAGEN
1503-00
470006-578EA 3.29 USD InStock
470006-578
Diffraction Grating
Educational Materials Physics Educational Materials Optics Learning Activities
A diffraction grating with 5400 lines per cm is mounted in a 5×5 cm mounting frame.
The diffraction grating exposed is one square inch.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty