Would you like to log in again?
Digital Photogate Free Fall Apparatus
Supplier: VWR International
GFF-AL PHGST08
470110-382EA 421.4 USD InStock
470110-382
Digital Photogate Free Fall Apparatus
Educational Materials Physics Educational Materials Mechanics Learning Activities
The Digital Photogate Free Fall Apparatus retains the precision of older models while eliminating the hassle of setting up heavy steel columns and adjusting spark recorders.
  • Ergonomic Design
  • Precise
  • Easy-To-Use
  • Steel Ball
  • Two Adjustable Photogates
The apparatus consists of a lightweight, extruded vertical column that carries a release electromagnet, two adjustable photogates, and a catching bag. The falling body is a 1.8 cm dia. steel ball.
The required Digital Timing System, sold separately, has several operating modes that allow different series of up to ten measurements to be made. These include the times of fall from rest to the first photogate, the times to fall between the two photogates, and the times to pass through each photogate. After the conclusion of a series, the apparatus displays the measured times. This comprehensive set of data not only allows a precise determination of the gravitational acceleration, but also permits verification of the equations of accelerated motion.
Delivery information: This item is not available for sale in Canada.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty