Would you like to log in again?
Dihybrid Cross
Supplier: VWR
470179-606KT 105 USD InStock
470179-606
Dihybrid Cross
Specimens Live Specimens Fungi - Living Material
Supplied as tube cultures. Comes with culture and handling instructions, as well as the manual Working with Fungi.
Eight strains: a1 (HAO), α1 (HBO), a2 (HAR), α2 (HBR), a3 (HAT), α3 (HBT), a4 (HART), α4 (HBRT).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty