Would you like to log in again?
Handheld Discharge Electrode
Supplier: SCIENCE INTERACTIVE GROUP
N-122
470005-530EA 158.45 USD InStock
470005-530
Handheld Discharge Electrode
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Handheld Discharge Electrodeeasily and safely discharges an electrostatic generator.
  • Polished Aluminum Sphere
  • Insulated Lucite Rod
  • Flexible Wire Lead
  • Generator Attaches to Wire Lead
  • Electrode Discharges an Electrostatic Generator
A polished aluminum sphere, 10.5 cm in diameter, is mounted on a 40 cm insulated Lucite rod.
A flexible wire lead has a banana plug at one end and a clip on the other, so it can attach to the base of the generator.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty