Would you like to log in again?
Ward's® Disinfectant Sensitivity Lab Activity
Supplier: Avantor
WARD470024-564
470105-820KT 165.09 USD InStock
470105-820 470024-564
Ward's® Disinfectant Sensitivity Lab Activity
Educational Classroom Kits and Activities
Test Effectiveness of Various Commercial Cleaners
Do disinfectant cleaners really work as advertised? Find out by testing five disinfectant samples against two common nonpathogenic bacterial strains. Students place disinfectant disks on a bacterial culture; if it is effective against the bacteria, a clear zone will appear around the disc. It comes with tube cultures of Bacillus subtilis and Escherichia coli, materials for six setups, teacher’s guide, and student copymaster. Note: Coupon included for perishable materials. Redeem by mail, fax, phone, or e-mail
Available to US customers only.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty