Would you like to log in again?
Ward's® DNA Extraction of Wheat Germ Lab Activity
Supplier: Avantor
470024-090KT 90.59 USD InStock
470024-090
Ward's® DNA Extraction of Wheat Germ Lab Activity
Educational Classroom Kits and Activities
Procedure Yields Large Amounts of Plant DNA
A knowledge of the properties and characteristics of DNA is crucial to gaining an understanding of molecular genetics and biotechnology. Using a special detergent, alcohol, and other materials, students will perform a series of procedures that will release and precipitate DNA from wheat germ cells where it can be spooled and examined for such properties as viscosity and elasticity. They will then compare the physical properties of the extracted DNA to the included macromolecular solution and note the similarities and differences. The whole procedure can be completed in one class period. You will get materials for 15 setups, a teacher’s guide, and student copymaster. Note: Coupon included for perishable materials. Redeem by mail, fax, phone, or mail.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty