Would you like to log in again?
DNA On A Chain Kit
Supplier: Ward's
WARD470012-514 605-BVP
470012-516EA 3.35 USD InStock
470012-516 470012-514
DNA On A Chain Kit
Educational Classroom Kits and Activities
It's a unique, engaging way of learning about DNA extraction.
  • Students Explore DNA Extraction
  • Instructions and Materials for 30 Students Included
  • Developed In Partnership with Brookhaven National Laboratory
  • Activities Features Real-World Applications
  • Time: 45 Minutes
Developed in partnership with Brookhaven National Laboratory, this DNA extraction lab involves a fun twist that both simplifies the process and helps guarantee understanding. Students swish a sports drink (not included) in their mouth to break open or lyse their own cheek cells.
This fluid is then added to a detergent containing sodium dodecyl sulfate (SDS) and heated to break down the fats and proteins of the cell membrane. Ethanol is gently added to the solution to reveal the DNA precipitate. Once isolated, students can display their own DNA using the colorful cord and microcentrifuge tubes included.
Ordering information: This kit includes plastic inoculating loops, a lab marker, nylon string, paper cups, detergent, 15 ml centrifuge tubes, microcentrifuge tubes, transfer pipets, and ethanol. Sports drinks, a hot-water bath or hot plate, graduated cylinders, ice, and a test tube rack are needed but not included.
Delivery information: Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty