Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
DNA Staining Kit
Supplier: LAB-AIDS INC (KITS) SE
170
470200-500KT 188 USD InStock
470200-500
DNA Staining Kit
Stains Electrophoresis Stains
Using the Feulgen procedure, students macerate, stain and mount specially grown and selected root tips included in the kit. Then, using a microscope, they examine and draw what they observe of the various stages of cell division.
Includes: 1 vial specially selected root tips; 2 drop controlled btls. each 1M hydrochloric acid, leucofuchsin dye, and sodium bisulfite solution; 1 drop controlled btl. dilute acetic acid; 30 plastic treating dishes; 30 transfer brushes; 30 microscope slides; 30 cover slips; 30 students worksheets and guides; 1 instruction manual with MSDS.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty