Would you like to log in again?
Dressing Stick for Grinders
Supplier: Saint-Gobain
61463610599
470022-256EA 49.65 USD InStock
470022-256
Dressing Stick for Grinders
Scissors
Extends Life of Diamond Cup Wheel.
A silicon carbide stick helps keep your grinder working smoothly. Size: 1/2 ×1/2×6".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty