Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ward's® Drosophila Culturing Set
Supplier: VWR
470180-400EA 18.95 USD InStock
470180-400
Ward's® Drosophila Culturing Set
Specimens Live Specimens Animals - Living Material
Readily Observed Strain Requires No Magnification
Use this unique culturing set as a convenient introduction to genetics; students can perform a simple monohybrid cross to yield a 3:1 winged: wingless ratio. Females have red eyes with yellow bodies, males have white eyes with gray bodies, making them easy to distinguish. Cultures supplied are yellow-forked-attached and white, and yellow-forked-attached and white apterous. The set comes with WARD’S Drosophila Culture Guide and cultures for up to 15 setups.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty