Would you like to log in again?
Ward's® Dual Power Supply
Supplier: Ward's
365110
470024-002EA 471.9 USD InStock
470024-002
Ward's® Dual Power Supply
Power Supplies
Feature-Rich Yet Economical for Educational Use
Save space in the laboratory with the small footprint of this unit. It allows you to run up to two electrophoresis chambers simultaneously.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty