Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ward's® Earth-Sun Motion Lab Activity
Supplier: Avantor
WARD470023-780
470328-414KT 86.19 USD InStock
470328-414 470023-780
Ward's® Earth-Sun Motion Lab Activity
Educational Classroom Kits and Activities
Tell Time from the Shadows Made by the Earth’s Rotation.
  • Create a primitive sundial
  • Observe Earth's rotation
  • Reusable
  • Aligned to NGSS standards (GRK-5/MS: ESS1)
Students construct shadow sticks, which functioned as an early sundial, to observe how changes in a shadow are related to the Earth’s rotation and are used to measure time. It comes with enough materials for 25 students, a teacher’s guide, and student copymasters.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty