Would you like to log in again?
Economy Spherical Mirror Set
Supplier: United Scientific Supplies
MCVSET
470111-310KT 26.55 USD InStock
470111-310
Economy Spherical Mirror Set
Mirrors
Put One in the Hands of Every Student in Your Class
  • Package of six
The mirrors are made of 3mm thick glass with back surface silvering. You get six spherical mirrors with 50 mm diameters. Three concave and three convex with three focal lengths each, 60, 100 and 200mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty