Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Elasticity Demonstrator
Supplier: Avantor
WARD470129-282
470129-282KT 79.99 USD InStock
470129-282
Elasticity Demonstrator
Educational Materials Physics Educational Materials Mechanics Learning Activities
Low cost easy to use lab for studying elastic and inelastic collisions.
It includes a curved aluminum track with base for attaching to a table or bench with a C-clamp; a block of wood; and two rubber balls which are identical in appearance but differ greatly in the degree to which they bounce (Happy/Sad balls).
The 'Happy' ball, which bounces, is made of neoprene rubber. The 'Sad' ball, which does not bounce, is of polynorbornene rubber.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty