Would you like to log in again?
Electric Sieve Shaker
Supplier: Humboldt
H4325
470018-706EA 1751.34 USD InStock
470018-706
Electric Sieve Shaker
Shakers and Mixers Sieve Shakers
Assure Accurate and Dependable Separations Every Time
Our electric sieve shaker is specially designed for both vertical and horizontal tapping action. Spring-loaded tie rods and a 30-minute, built-in timer offer a steady, regulated shaking action. This unit has a larger capacity than most models, accommodating up to nine standard 8" sieves, not included, as well as a bottom pan and lid. A built-in adapter and convenient lid lock allow the unit to hold fewer sieves, or to hold smaller 3" or 6" diameter sieves. It is powered by a quiet-running, beltdriven, 1/4 hp motor. 115 VAC, with a 5 1/2' cord; 60 Hz. Size 14"L x 12"W x 36"H; weight: 65 lbs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty