Would you like to log in again?
Electric Timer for Plant Centers
Supplier: BOSTWICK-BRAUN COMPANY
580759
470103-648EA 62.35 USD InStock
470103-648
Electric Timer for Plant Centers
Timers Digital Timers
Make sure your plants get the right amount of light with this 24-hour timer, complete with two on and off settings per day for flexibility in programming.
The all-weather plastic case never rusts or corrodes, and the unit can be used either indoors or outdoors in temperatures from ‒40 to 49 °C.
Delivery information: A three-foot grounded cord, designed for 120 V, AC service, is supplied.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty