Would you like to log in again?
Electrical Field Apparatus
Supplier: VWR International
PH0889
470120-702EA 45.8 USD InStock
470120-702
Electrical Field Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Directly View Electric Field Patterns
Much in the same way magnetic fields are demonstrated with iron fillings, this apparatus allows electric fields to be viewed. For use on a desktop or overhead projector, a plastic dish and base features terminals with a thumbscrew system, which permits fine height adjustment of the electrodes. Six electrodes are supplied — two point source, two line source, and two circular rings. The dish is filled to a depth of 6 - 8 mm with liquid paraffin, or other non-conducting fluid (these are not included). A small amount of semolina (not include) is sprinkled evenly on the surface of the liquid. When the terminals are connected to the power supply field patterns are formed in the semolina.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty