Would you like to log in again?
Vernier® Electrostatics Kit
Supplier: Vernier
ESK-CRG
470017-514EA 128.99 USD InStock
470017-514
Vernier® Electrostatics Kit
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Inclusive package for inclusive study.
  • Using Faraday’s Ice Pail
  • Production of charge by friction
  • Transfer of charge by contact
  • Charging by Induction
  • Quantitative and qualitative measurement of charge
An accompaniment to the Vernier Charge sensor, this kit enables the inquiry and examination into a wide range of experiments in electrostatics. Measure charge in a quantitative and qualitative manner, and see how friction, induction, and the contact of two objects can vastly affect its charge. The kit includes everything you need for deep exploration- a Faraday pail and cage, grounding wires and plane, a wrist strap, a proof plane, and the following charge producers: wool, vinyl, a PVC rod, a nylon rod, and cotton cloth.
For educational use only
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty