Would you like to log in again?
Ward's® Live Elodea densa Aquatic Plants
Supplier: VWR
Ward's
WARD470180-036 WARD470198-312 WARD470175-814 WARD470176-598 WARD470180-034 WARD470180-038
470180-034PK 13.99 USD InStock
470180-034 470180-036 470175-814 470180-038 470176-598 470198-312
Ward's® Live Elodea densa Aquatic Plants
Specimens Live Specimens Plants - Living Material
An excellent aquarium oxygenator, Elodea densa is great for teaching plant respiration, photosynthesis, and plasmolysis.
  • Sprigs can be rooted in substrate for aquarium use
  • Tips have fresh growth to demonstrate plasmolysis, streaming protoplasm and osmosis
Elodea densais an excellent alternative if Elodea canadensis or Egeria najas are restricted in your state.
Tips are not recommended for aquarium use. Instead sprigs can be rooted in substrate, or floated on water as a food source or hiding place for fish and snails.
Ordering information: May be restricted in some states, check with your state's Department of Agricutlrure for restrictions. Elodea canadensis or Egeria najas are also available as alternatives.
Delivery information: These specimens are perishable and will ship via 2-day delivery. Please provide a requested delivery date.
Packaging: Plants are provided in waterproof, plastic bags that keep plants both fresh and healthy. Care instructions available online.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty