Would you like to log in again?
Epididymis, Primate Slide
Supplier: Avantor
WARD470183-210
470183-210EA 7.45 USD InStock
470183-210
Epididymis, Primate Slide
Slides Prepared Slides
Male Reproductive System Component
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Primate. Pseudostratified epithelial lining of tubules, connective tissue, blood vessels, and masses of spermatozoa.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty