Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ward's® Exploring Deposition of Sediments Lab Activity
Supplier: Avantor
WARD470012-448
470012-448KT 718.99 USD InStock
470012-448
Ward's® Exploring Deposition of Sediments Lab Activity
Educational Classroom Kits and Activities
Study Sediment Deposition, Transport, and Distribution
  • See factors involved with sediment deposition
  • Includes sedimentation tube and multiple sediment sizes
Students can see how particle size, shape, and density affect the settling rate of various materials by testing how quickly they settle in the specially constructed four-foot-long sedimentation tube. Through observation, and recording and graphing the settling rates, students compare the results and draw conclusions about sediment deposition in nature. It comes with a teacher’s guide and student copymasters.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty