Would you like to log in again?
Ward's® Exploring Physical Properties of Minerals Lab Activity
Supplier: Avantor
WARD470002-460
470328-402KT 298.29 USD InStock
470328-402 470002-460
Ward's® Exploring Physical Properties of Minerals Lab Activity
Educational Classroom Kits and Activities
Students Master Investigative Skills by Testing Mineral Properties.
  • Teach students about properties of minerals
  • Teacher and student guides included
  • Aligned to NGSS standards (MS/HS: ESS2, ESS3)
Eight common minerals are supplied to test hardness, color, streak, luster, specific gravity, and other characteristics. Each sample has been carefully selected to ensure that students can readily recognize key physical properties and achieve consistent test results. Each kit contains a streak plate, copper strip, glass plate, nail, magnifier, and graduated cylinder for physical tests, as well as a teacher’s guide and worksheet copymasters.
Minerals: calcite, feldspar, galena, graphite, hematite, muscovite, pyrite, and quartz.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty