Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EZ Fill Skinny Container

Supplier: Impact Products
7572-90
470104-702EA 55.55 USD InStock
470104-702
EZ Fill Skinny Container
Bottles Carboys
Heavy-duty container with a 63 mm angled-front opening for refilling.

Includes #7577 faucet for dispensing and 38 mm cap for top vent.

Certifications: Complies with Federal regulations for food contact.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty