Would you like to log in again?
Fieldmaster Lake Bottom Sampling Kit
Supplier: SCIENCE INTERACTIVE GROUP
623-7210
470021-250EA 460.9 USD InStock
470021-250
Fieldmaster Lake Bottom Sampling Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Featuring Durable, Reusable Equipment
Explore the contents of ponds, lakes, and other bodies of water through examination of samples collected at the water’s hard or soft bottom. This kit features sturdy, student friendly supplies, including the Mighty Grab™, which needs no messenger and can be operated either by hand line or with the included sampling pole. A wash bucket with 504 micron stainless steel mesh sorts samples to the same standards used by the professionals. An armored glass thermometer (-40°F to 120°F) is also provided for taking samples of air and water. Take samples back to the classroom and lab with durable polypropylene wide-mouth sample bottles, designed with a square shape that won’t tip or spill. Comprehensive teacher instructions included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty