Would you like to log in again?
Fieldmaster Stream Sampling Kit
Supplier: SCIENCE INTERACTIVE GROUP
623-7211
470021-252EA 432.5 USD InStock
470021-252
Fieldmaster Stream Sampling Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Economical and Easy to Use
Obtaining samples from flowing water for further study with the contents of this collecting kit. Several students can gather samples simultaneously with the included equipment designed to not require extensive training. Six nets and six sample jars are included. The D-frame net is lightweight and practical, with a 30" handle for easy handling. The wide-mouthed polypropylene sample jars hold up to 500mL and are square in shape so they won’t tip or spill. The provided turbidity tube is student-sized at 60cm in length and functions like a miniature secchi disk to measure water clarity. An armored glass thermometer (-40°F to 120°F) is also supplied for for taking accurate, relevant temperatures of air and water. Six 500 mL plastic wash bottles are also provided for rinsing down equipment in the field. Includes comprehensive teacher instructions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty