Would you like to log in again?
Fieldmaster Water Sampling Field Kit
Supplier: SCIENCE INTERACTIVE GROUP
623-7215
470021-248EA 536 USD InStock
470021-248
Fieldmaster Water Sampling Field Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
Comprehensive, Cost-Effective Collecting Kit
  • Collect plankton samples
  • Measure water quality parameters
  • Includes a rugged carrying case for equipment
Everything your students need for sampling water for plankton organisms, to determine the water transparency index, and to measure water depth up to twenty meters is included. The kit features a clear polycarbonate water sampler, Secchi disk and line on float, 5" diameter mouth plankton net in 153 micron mesh with an attached 125 mL sample bottle, and thermometer (-40°F to 120°F) — all in a plastic carry case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty