Would you like to log in again?
Fieldmaster Plankton Field Kit
Supplier: SCIENCE INTERACTIVE GROUP
623-7212
470021-254EA 368 USD InStock
470021-254
Fieldmaster Plankton Field Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
For Samples of Phyto and Zooplankton
  • Collect plankton samples
  • Measure water quality parameters
  • Includes instructions
Uniquely designed for student sample collecting, the supplies in this kit are durably constructed and ideal for transporting samples back to the classroom or lab. The basic water bottle provided is constructed of PVC with soft rubber seals and 20m of line on a styrofoam float that will bob to the surface if dropped. The plankton net is constructed of genuine Nitex® 80 micron mesh with a stainless steel ring and bridle for attaching line. 250mL sample bottles and 125mL bottles fit into the plankton net with the included adaptor and allow for multiple samples. The secchi disk meets industry standards, determines turbidity in shallow waters, and comes with attached line on a winding float. A 500mL wash bottle and 3/4" long thermometers are also included. Comprehensive teacher instructions provided.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty