Would you like to log in again?
Ward's® Fish Scale Set
Supplier: VWR
WARD470000-984
470000-984KT 51.99 USD InStock
470000-984
Ward's® Fish Scale Set
Specimens Preserved Specimens Preserved Animal Materials
Formalin Preserved
Examine the different types of external structure unique to fish. Contents: ten each of placoid, ganoid, ctenoid, and cycloid scales. The set of four is supplied in separate jars.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty