Would you like to log in again?
WARD’S “Fool’s Gold” Prospector Lab Activity
Supplier: Avantor
WARD470025-216
470025-216EA 43.99 USD InStock
470025-216
WARD’S “Fool’s Gold” Prospector Lab Activity
Educational Classroom Kits and Activities
Mine for “Gold” and learn about minerals at the same time.
There’ll be a gold rush in your classroom when your students use our exclusive lab activity. The specially prepared sediment provided contains fool’s gold fines in a bulk matrix. Students will use panning techniques to separate the “gold” from the sediment. In the process, they learn about mineral density and heavy mineral prospecting. It comes with a 1/2 gallon (approximately 6 lbs.) of sediment and instructions on panning. A gold pan, required, is available separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty