Would you like to log in again?
Forensic Foot Powder Mystery Kit
Supplier: Avantor
WARD470007-250
470007-250KT 202.99 USD InStock
470007-250
Forensic Foot Powder Mystery Kit
Educational Classroom Kits and Activities
Use forensic procedures to solve the case of the stinky feet.
  • Grades 6-9
  • Real world case based scenario
  • Estimated class/lab time required is 90 minutes over 2 days
Challenge students to crack the case using actual forensic science techniques. After testing various powders in higher temperatures and three different liquids, students record their observations and compare them to a sample from the crime scene. Developed by teacher Molly Phelan, this unique activity includes materials for eight setups.
Materials included:
5 powder samples
Spot plates
Test tube clamp
Dropper Bottle
Hand lens
Laboratory Scoop
Vinegar
Iodine
Foil
Spoons
Toothpicks
Instructions
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty