Would you like to log in again?
Freeze-Dried Sheep Brain
Supplier: VWR
WARD470027-886 WARD470027-884
470027-886PK 174.99 USD InStock
470027-886 470027-884
Freeze-Dried Sheep Brain
Specimens Preserved Specimens Preserved Animal Materials
Freeze-dried sheep brain. Available in vacuum packs.
  • Mammalian anatomy
  • Economical
Preserved without chemicals or fluids, this brain can be used for external study and dissection. The specimen can be rehydrated by adding a 20% alcohol solution.
Note: Ward's® preserved sheep materials is a byproduct of the food industry. Sheep are not raised specifically for dissection. These animals are recycled" for educational use."
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty