Would you like to log in again?
Freeze-Dried Sheep Lungs
Supplier: VWR
WARD470027-874
470027-874EA 140 USD InStock
470027-874
Freeze-Dried Sheep Lungs
Specimens Preserved Specimens Preserved Animal Materials
Freeze-Dried Preserved.
Completely dry, so they can be sectioned with a scalpel to show the internal structure, these lungs can be displayed easily, without fear of chemical or fluid leaks. To enhance their durability, they have been treated with a clear plastic sealant. Note: Not intended for rehydration.
Note: Ward's® preserved sheep materials is a byproduct of the food industry. Sheep are not raised specifically for dissection. These animals are recycled" for educational use."
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty