Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Ward's® Fruit Fly Genetics Demonstration
Supplier: Avantor
470030-324KT 59.69 USD InStock
470030-324
Ward's® Fruit Fly Genetics Demonstration
Educational Classroom Kits and Activities
Demonstrate a simple monohybrid cross using the fruit fly Drosophila melanogaster
  • No microscope needed to view the offspring from this cross
  • Mutations are easily visible
The cross between winged and wingless fruit flies yields a 3:1 winged:wingless ratio.
Males will have white eyes and gray bodies; females will have red eyes and yellow bodies.
All necessary materials are provided, including a hand lens with which students can easily distinguish the trait, as well as males and females.
Kit contains:
Ice pack and anesthetizer to sedate flies
Empty vials and plugs
Instructions and a coupon redeemable for live flies.
Ordering information: This item contains a copupon for live or perishable materials. At checkout, please provide the date you wish the perishable components to arrive.
Delivery information: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground.
Perishable components will ship via overnight or second day delivery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty