Would you like to log in again?
Ward's® Fungi Classification Set
Supplier: Avantor
WARD470347-316 WARD470177-264
470177-264EA 73 USD InStock
470177-264 470347-316
Ward's® Fungi Classification Set
Educational Materials Biology Educational Materials Classification Learning Activities
Representatives from the four major fungal groups.
  • Aligned to NGSS standards (MS/HS: LS1, LS2)
Six cultures, all grown in our own labs and sealed at the optimal point of fungal growth, clearly illustrate the structural differences between the four fungal groups. Demonstration plate cultures: Schizophyllum commune (Basidiomycete); Aspergillus niger, Penicillium notatum (Deuteromycete); Mucor heimalis, Rhizopus stolonifer (Zygomycete); and Neurospora crassa (Ascomycete). The set of six comes with information on fungal structure and suggested activities, as well as the Working with Fungi manual.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.
Delivery information: Allow two weeks for delivery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty