Would you like to log in again?
General Animal Slide Set
Supplier: Avantor
WARD470183-506
470183-506KT 199.99 USD InStock
470183-506
General Animal Slide Set
Slides Prepared Slides
Illustrate Comparative Differences Between Multicellular Organisms in Major Phyla
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Ordering information: Slide Set contents include Poriferans (1 slide), Coelenterates (2 slides), Nematodes (1 slide), Platyhelminthes (1 slide), Annelids (1 slide), Crustaceans (1 slide), Arthropods, insects (4 slides), Echinoderms (1 slide), Molluscs (1 slide), Vertebrates, fish (1 slide), Vertebrates, amphibians (4 slides), Vertebrates, birds (3 slides), and Chordate tissues (4 slides). Note: Please call Customer Service for a complete listing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty