Would you like to log in again?
General Epithelial Tissues Slide Set
Supplier: Avantor
WARD470183-586
470183-586CS 816 USD InStock
470183-586
General Epithelial Tissues Slide Set
Slides Prepared Slides
Illustrates the Types and Functions of Epithelia
  • All identifying features are clearly distinguishable
  • Rigorous quality control standards
  • In-house experts available to answer your questions
Learn about the many types of epithilia and their form and function. All slides are prepared as sections unless otherwise noted.
Ordering information: Slide Set includes: Literature; Squamous, frog skin (wm); Squamous, intracellular bridges; Simple cuboidal, kidney; Simple columnar, intestine; Pseudostratified ciliated columnar, respiratory tissue; Stratified squamous, esophagus; Transitional, stratified cuboidal; excretory; Neuro-epithelium, taste bud; Ciliated, isolated; Pigmented skin; Prostate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty